Pharmaceutical Excipients FT-IR Spectral Library
Thermo Scientific™
Pharmaceutical Excipients FT-IR Spectral Library
Get high quality FT-IR spectra of the most common pharmaceutical excipients in the Thermo Scientific™ Pharmaceutical Excipients FT-IR Spectral Library.
Have Questions?
Catalog NumberType
834-019201Pharmaceutical Excipients FT-IR Spectral Library
Catalog number 834-019201
Price (INR)
-
Type:
Pharmaceutical Excipients FT-IR Spectral Library
Get high quality FT-IR spectra of the most common pharmaceutical excipients in the Thermo Scientific™ Pharmaceutical Excipients FT-IR Spectral Library, designed for laboratories studying pharmaceutical formulations using FT-IR spectroscopy.
Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.
This library provides high quality FT-IR spectra of the most common pharmaceutical excipients.
  • Consists of 300 spectra collected using a Thermo Scientific FT-IR spectrometer
  • All materials were tested via current United States Pharmacopoeia compendium methodology for identity and purity—only materials that passed these methods were used to create this library
  • Data format: High Resolution (HR)
Includes:
Textbook explaining spectroscopy techniques and the spectra of pharmaceutical excipients for reference use.
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypePharmaceutical Excipients FT-IR Spectral Library
Unit SizeEach