Near IR Pharma Reference Database Spectral Library
Thermo Scientific™
Near IR Pharma Reference Database Spectral Library
FT-NIR spectral library collection of active and inactive pharmaceutical substances.
Have Questions?
Catalog NumberType
834-024301Near IR Pharma Reference Database Spectral Library
Catalog number 834-024301
Price (INR)
-
Type:
Near IR Pharma Reference Database Spectral Library
Thermo Scientific™ Near IR Pharma Reference Database Spectral Library offers an FT-NIR spectral library collection of active and inactive pharmaceutical substances.
This high resolution library can be used to identify unknown samples, as a reference when setting up qualitative NIR methods, or as a comparison against known sample spectra.
  • Consists of spectra of 385 pharmaceutical compounds including both active and inactive substances
  • Solid samples collected in reflectance using a SabIR™ fiber optics probe
  • Liquid samples collected in transmittance using a 2mm quartz cell
  • Data format: High Resolution
Specifications
For Use With (Application)FT-IR
TypeNear IR Pharma Reference Database Spectral Library
Unit SizeEach