Inorganics FT-IR Spectral Library
Thermo Scientific™
Inorganics FT-IR Spectral Library
Provides spectra of inorganic and related compounds from around the world.
Have Questions?
Catalog NumberType
834-025800Inorganics FT-IR Spectral Library
Catalog number 834-025800
Price (INR)
-
Type:
Inorganics FT-IR Spectral Library
Thermo Scientific™ Inorganics FT-IR Spectral Library provides spectra of inorganic and related compounds from around the world.
This library consists of minerals, boron compounds, general inorganic substances and inorganic materials of commercial interest.
  • Contains 1,803 total spectra, including 600 minerals, 296 boron compounds, 698 other inorganic substances and 211 commercial materials
  • Commercial materials include cements, ceramics, porcelain, clay, glass and other raw materials
Specifications
For Use With (Application)FT-IR
TypeInorganics FT-IR Spectral Library
Unit SizeEach