Cayman Chemical™ Raman Library
Thermo Scientific™
Cayman Chemical™ Raman Library
Advance your research with this Raman library containing spectra of synthetic psychoactive substances such as synthetic cannabinoids, synthetic cathinones, synthetic piperazines, and tryptamines.
Have Questions?
Catalog NumberFor Use With (Equipment)
834-105000DXR Raman microscope; DXRxi Raman imaging microscope; DXR SmartRaman spectrometer
Catalog number 834-105000
Price (INR)
-
For Use With (Equipment):
DXR Raman microscope; DXRxi Raman imaging microscope; DXR SmartRaman spectrometer
The Cayman Chemical™ Raman reference library contains spectra of synthetic psychoactive substances based on pure reference material obtained from Cayman Chemicals.
This 187 spectra library include synthetic cannabinoids, synthetic cathinones, synthetic piperazines, and tryptamines, which are the basis for newer drugs of abuse such as spice, bath salts, legal ecstasy, and novel psychoactive drugs.
For use with: Thermo Scientific™ DXR™ Raman microscope; Thermo Scientific™ DXR™xi Raman imaging microscope; Thermo Scientific™ DXR™ SmartRaman spectrometer
Specifications
DescriptionCayman Chemical Raman Library
For Use With (Equipment)DXR Raman microscope; DXRxi Raman imaging microscope; DXR SmartRaman spectrometer
Unit SizeEach