Nicolet Narcotics FTIR Spectral Library
Nicolet Narcotics FTIR Spectral Library
Thermo Scientific™
Nicolet Narcotics FTIR Spectral Library
The Thermo Scientific™ Nicolet™ Narcotics FTIR Spectral Library contains a total of 883 spectra of new psychoactive substances including fentanyl.
Have Questions?
Catalog NumberFor Use With (Equipment)
834-138200FTIR
Catalog number 834-138200
Price (INR)
-
Request A Quote
For Use With (Equipment):
FTIR
Take the guesswork out of your spectral analysis with the Thermo Scientific™ Nicolet™ Narcotics FTIR Spectral Library. This comprehensive library features fully-reviewed, high-quality spectroscopy databases, enabling compound identification for forensic applications with a total of 883 spectra of new psychoactive substances (NPS) including 190 fentanyl analogues.
Analysis of narcotic samples can be greatly enhanced with the help of spectral libraries. Data can be analyzed and summarized reliably using vast libraries of high-resolution spectra. This database includes a total of 883 spectra of new psychoactive substances (NPS) including 190 fentanyl analogues.
About Spectral Libraries:
  • Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox.
  • Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FTIR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification—or at least classification—of the material.
  • Our extensive series of spectral libraries include information specific to application, industry or spectral technique. Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
DescriptionNicolet Narcotics FTIR Spectral Library
For Use With (Equipment)FTIR
Unit SizeEach