Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer.
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
Type
840-214900
Evolution Spectrophotometers
7-Cell Rotary Changer
10011401
Smart Rotary Cell Changer Base
Temperature Probe
10010110
Smart Rotary Cell Changer Base
Spare 7-cell Rotary Changer
3 Options
Catalog number 840-214900
Price (INR)
-
For Use With (Equipment):
Evolution Spectrophotometers
Type:
7-Cell Rotary Changer
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer. Combined with a liquid recirculator, this cell changer provides control of sample temperature for up to 7 samples or standards.
Easy-to-use sample positioning accessory for use with Thermo Scientific™ Evolution™ One/One Plus and Evolution™ Pro spectrophotometers.
Accessory control fully integrated in Thermo Scientific™ INSIGHT™ Pro software.
Automatic software recognition, smart alignment and serial number reporting
Includes carousel and motor base.
When placed in both sample and reference beams (Evolution 300 only), this cell changer allows comparison of up to 7 samples with 7 unique references.
Monitor the temperature in a cell with Temperature probe accessory for Smart accessories, catalog number 0011401.
When deploying this accessory in a regulated or QC environment, a full-system qualification package is available to assure compliance of instrument and accessory as a complete unit.
Specifications
CompatibilityCompatible with temperature probe accessory for Smart accessories, catalog#10011401
Configuration DetailsEvolution One/One Plus- sample position only, Evolution Pro- sample and reference positions
For Use With (Equipment)Evolution Spectrophotometers
IncludesCarousel and motor base
ThermostattingThermostatting with flowing liquid from an external temperature controlled recirculator