NanoDrop™ Cuvette Qualification Filter Kit
NanoDrop™ Cuvette Qualification Filter Kit
NanoDrop™ Cuvette Qualification Filter Kit
NanoDrop™ Cuvette Qualification Filter Kit
Thermo Scientific™
NanoDrop™ Cuvette Qualification Filter Kit
The Cuvette Qualification Filter Kit for the cuvette based Thermo Scientfic™ NanoDrop™ OneC and NanoDrop™ 2000c Spectrophotometers is an essential tool for ensuring the optimal performance of your spectrophotometers. This kit provides you with the necessary filters for accurate cuvette qualification testing.
Have Questions?
Catalog NumberFor Use With (Equipment)
840-257300NanoDrop OneC and NanoDrop 2000C Spectrophotometers
Catalog number 840-257300
Price (INR)
167,013.00
Each
Add to cart
For Use With (Equipment):
NanoDrop OneC and NanoDrop 2000C Spectrophotometers
Price (INR)
167,013.00
Each
Add to cart
The Cuvette Qualification Filter Kit for the cuvette based Thermo Scientfic™ NanoDrop™ OneC and NanoDrop™ 2000c Spectrophotometers is an essential tool for ensuring the optimal performance of your spectrophotometers. This kit provides you with the necessary filters for accurate cuvette qualification testing.
Includes: 1 x 10%T Filter (Calibrated), 1 x 50%T (Calibrated), and a certificate with traceability and NVLAP 17025, Lab Code 200955, accreditation.
Specifications
DescriptionQualification Filter Kit for NanoDrop OneC and NanoDrop 2000C Spectrophotometers
For Use With (Equipment)NanoDrop OneC and NanoDrop 2000C Spectrophotometers
Includes1 x Filter holder blank, 1 x 10%T Filter, Calibrated, 1 x 50%T Filter, Calibrated, Certificate with traceability and NVLAP 17025, Lab Code 200955, accreditation
Unit SizeEach