GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
Thermo Scientific™

GENESYS™ 30 Visible Spectrophotometer Accessories

Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
840-278400Dust Cover
085-820200USB Cable
840-278700Tungsten Halogen Lamp
840-278500Replacement Compartment Lid
Catalog number 840-278400
Price (INR)
-
Request A Quote
Type:
Dust Cover
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible In addition to valuable built-in features such as the high resolution color display, large sample compartment, and data export options, a complete selection of accessories are available for the the GENESYS 30 spectrophotometer to help meet the needs of students, educators and lab technicians for accurate quantitative analysis.

  • Holds one rectangular longpath cell 20 - 100mm pathlength 
  • Comes assembled in a removable sample compartment liner
  • Allows for flexibility between samples
Specifications
For Use With (Application)Keep dust and spills off your spectrophotometer when it is not in use
For Use With (Equipment)GENESYS 30 Visible Spectrophotometer
Includes1 dust cover
Storage RequirementsAmbient
Warranty2 Years
TypeDust Cover
Unit SizeEach