GENESYS Fiber Optic Probes
Thermo Scientific™
GENESYS Fiber Optic Probes
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
840-305700GENESYS 150/180 SpectrophotometerFiber Optic Dip Probe System
840-305000GENESYS 150, GENESYS 180, Evolution One Plus, and Evolution Pro spectrophtometersFiber Optic Dip Probe, 10 mm
840-305800GENESYS 150/180 SpectrophotometerFiber Optic Coupler
840-307400GENESYS 150, GENESYS 180, Evolution One Plus, and Evolution Pro spectrophtometersTip for Dip Probe, 10 mm
Catalog number 840-305700
Price (INR)
-
For Use With (Equipment):
GENESYS 150/180 Spectrophotometer
Type:
Fiber Optic Dip Probe System
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Preserve sample integrity while minimizing sample carryover.
Specifications
Diameter (Metric)6 mm
For Use With (Equipment)GENESYS 150/180 Spectrophotometer
TypeFiber Optic Dip Probe System
Unit SizeEach