NanoDrop™ Eight Pipette Guide
NanoDrop™ Eight Pipette Guide
NanoDrop™ Eight Pipette Guide
Thermo Scientific™
NanoDrop™ Eight Pipette Guide
The Thermo Scientific™ NanoDrop™ Eight Pipette guide allows you to comfortably and accurately pipette samples onto all eight wells of the instrument using an 8-channel precision pipettor.
Have Questions?
Catalog NumberFor Use With (Equipment)
840-343600NanoDrop Eight Spectrophotometers
Catalog number 840-343600
Price (INR)
5,847.00
Online offer
Ends: 31-Mar-2026
9,744.00
Save 3,897.00 (40%)
Each
For Use With (Equipment):
NanoDrop Eight Spectrophotometers
Price (INR)
5,847.00
Online offer
Ends: 31-Mar-2026
9,744.00
Save 3,897.00 (40%)
Each
The Thermo Scientific™ NanoDrop™ Eight Pipette guide allows you to comfortably and accurately pipette samples onto all eight wells of the instrument using an 8-channel precision pipettor.
The pipette guide is magnetic and movable from side-to-side allowing left- and right-handed users to quickly and comfortably dispense samples
Specifications
DescriptionNanoDrop Eight Pipette Guide
For Use With (Equipment)NanoDrop Eight Spectrophotometers
Unit SizeEach
Frequently asked questions (FAQs)
Which is the best DNA or RNA quantification instrument?
All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
What is the difference between UV and fluorescence DNA quantification? Which DNA quantification technology should I choose?
UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity‒plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.