NanoDrop™ Eight IQ/OQ Kits
NanoDrop™ Eight IQ/OQ Kits
Actual product may vary
Thermo Scientific™
NanoDrop™ Eight IQ/OQ Kits
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ). Certified standards for verifying instrument performance to factory specifications are available separately.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
840-371200ND Eight Spec IQ/OQ – US
840-371300ND Eight Spec IQ/OQ – A4
840-371400ND Eight Spec IQ/OQ Requal
Catalog number 840-371200
Price (INR)
104,705.00
Online offer
Ends: 31-Mar-2026
174,507.00
Save 69,802.00 (40%)
Each
Add to cart
Type:
ND Eight Spec IQ/OQ – US
Price (INR)
104,705.00
Online offer
Ends: 31-Mar-2026
174,507.00
Save 69,802.00 (40%)
Each
Add to cart
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ). Certified standards for verifying instrument performance to factory specifications are available separately.
IQ/OQ Document Kits
Ensures your instrument is installed correctly and working within manufacturer’s specifications.
Includes a binder, paper (A4 or US), qualification labels, USB stick with
IQ/OQ procedure, CHEM-PR1-KIT, and CHEM-PV-8.
IQ/OQ Requalification Document Kit
Helps requalify your instrument for use on a routine schedule (e.g. annually) or after some event (e.g. service).
IQ/OQ requalification kit which includes a USB stick with IQ/OQ procedure, CHEM-PR1-KIT, and CHEM-PV-8. Diagnostic reagents also sold seperately.
Specifications
For Use With (Equipment)NanoDrop Eight Spectrophotometers
IncludesQualification Kit includes: IQ/OQ procedure for NanoDrop Eight Spectrophotometers, Qualification Binder Kit (US Size), CHEM-PR1-KIT, CHEM-PV-8
TypeND Eight Spec IQ/OQ – US
Unit SizeEach
Frequently asked questions (FAQs)
Which is the best DNA or RNA quantification instrument?
All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
What is the difference between UV and fluorescence DNA quantification? Which DNA quantification technology should I choose?
UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity‒plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.