Ceramic Purged Optical Path (POP) Nozzles for iCAP™ 7000 Series ICP-OES Systems
Thermo Scientific™
Ceramic Purged Optical Path (POP) Nozzles for iCAP™ 7000 Series ICP-OES Systems
For quality you can reply on, use replaceable Thermo ScientificTM Ceramic POP Nozzles for the advanced iCAPTM 7000 Series ICP-OES system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Includes
842312051161iCAP 7000 SeriesRadial Ceramic POP Nozzle
842315550541iCAP 6200/7200Axl Ceramic POP Nozzle (40 mm)
842312051261iCAP 7400/7600Axial Ceramic POP Nozzle
Catalog number 842312051161
Price (INR)
-
For Use With (Equipment):
iCAP 7000 Series
Includes:
Radial Ceramic POP Nozzle
For quality you can reply on, use replaceable Thermo Scientific™ Ceramic Purged Optical Path (POP) Nozzles for the Thermo Scientific™ iCAP™ 7000 Series ICP-OES Systems. Ceramic POP Nozzles form the purgeable interface between the plasma and optical system.
Compatibility
Ceramic POP Nozzles can be used with the iCAP 6000 Series ICP-OES and the iCAP 7000 Series ICP-OES instruments.
Specifications
IncludesRadial Ceramic POP Nozzle
For Use With (Equipment)iCAP 7000 Series
TypeCeramic POP Nozzle
Unit SizeEach