Search
| Catalog Number | Type |
|---|---|
| 842312051961 | Ceramic Center Tube, 2.0 mm |
| BRE0019895 | Quartz Center Tube, 2.0 mm |
| 842312051951 | Quartz Center Tube, 1.5 mm |
| 842312051941 | Quartz Center Tube, 1.0 mm |
| BRE0030178 | Ceramic Center Tube, 1.0 mm |
Available EMT center tubes include:
All EMT center tubes are compatible with Thermo Scientific™ iCAP™ 7000 Series and iCAP™ PRO ICP-OES systems.