Standard Argon Gas Filter Kit for the iCAP™ Series ICP-OES Systems
Thermo Scientific™
Standard Argon Gas Filter Kit for the iCAP™ Series ICP-OES Systems
Get everything needed to maintain the gas filtration system in our advanced ICP-OES systems in this useful Thermo Scientific™ Standard Argon Gas Filter Kit.
Have Questions?
Catalog NumberType
842312052121Argon Gas Filter
Catalog number 842312052121
Price (INR)
-
Type:
Argon Gas Filter
Enhance and optimize system performance by ensuring gas filters are replaced regularly to prevent contamination. This useful Thermo Scientific™ Standard Argon Gas Filter Kit for the Thermo Scientific™ iCAP™ Series ICP-OES systems includes everything required to maintain the argon gas supply filtration system in our advanced iCAP Series ICP-OES systems.
Specifications
For Use With (Equipment)iCAP Series ICP-OES Systems
TypeArgon Gas Filter
Unit SizeEach