Ceramic D-Torch Kits for the iCAP™ Series ICP-OES Systems
Actual product may vary
Ceramic D-Torch Kits for the iCAP™ Series ICP-OES Systems
Ceramic D-Torch Kits for the iCAP™ Series ICP-OES Systems
Thermo Scientific™
Ceramic D-Torch Kits for the iCAP™ Series ICP-OES Systems
Prevent devitrification and extend torch lifespan with the demountable Thermo Scientific™ Ceramic D-Torch for ICP-OES analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesFor Use With (Equipment)
842312052201Radial Ceramic D-Torch KitiCAP 7000/PRO Series
842312052202Duo Ceramic D-Torch KitiCAP 7000 Series
BRE0024347Duo Ceramic D-Torch KitiCAP PRO Series
Catalog number 842312052201
Price (INR)
-
Includes:
Radial Ceramic D-Torch Kit
For Use With (Equipment):
iCAP 7000/PRO Series
Reduce the cost of consumables traditionally associated with ICP-OES analysis using these demountable Thermo Scientific™ Ceramic D-Torch Kits for Thermo Scientific™ iCAP™ Series ICP-OES systems in place of a conventional quartz torch. These torch kits help prevent potential devitrification and significantly extend torch lifespan.
Enhanced robustness for difficult matricesDedicated versions of the Ceramic D-Torch are available for radial and duo view iCAP Series ICP-OES systems.
  • Radial Ceramic D-Torch Kit for radial-only systems
  • Duo Ceramic D-Torch Kit for axial and radial (duo) systems
Specifications
IncludesRadial Ceramic D-Torch Kit
For Use With (Equipment)iCAP 7000/PRO Series
TypeCeramic D-Torch Kit
Unit SizeEach