Outer Tubes for Ceramic D-Torches for iCAP™ Series ICP-OES Systems
Actual product may vary
Thermo Scientific™
Outer Tubes for Ceramic D-Torches for iCAP™ Series ICP-OES Systems
Improve the durability of your plasma torch using a ceramic D-Torch with a ceramic or quartz outer tube for Thermo Scientific™ iCAP™ Series ICP-OES systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
842312052207Quartz Outer TubeDuo D Torch - iCAP 7000
842312052205Ceramic Outer TubeDuo D Torch - iCAP 7000
BRE0018578Ceramic Outer TubeDuo D-Torch - iCAP PRO
842312052208Quartz Outer TubeRadial D Torch-iCAP 7000/PRO
842312052206Ceramic Outer TubeRadial D Torch-iCAP 7000/PRO
Catalog number 842312052207
Price (INR)
-
Type:
Quartz Outer Tube
For Use With (Equipment):
Duo D Torch - iCAP 7000
Improve ICP-OES plasma torch lifespan by switching to a ceramic D-torch. Replacement Thermo Scientific™ Outer Tubes for Thermo Scientific ICP-OES Ceramic D-Torches are made from a durable, ceramic material derived from silicon nitride, which is resistant to chemical degradation. These properties are maintained at high temperatures and help prevent the potential devitrification associated with traditional quartz torches. When needed, quartz replacement outer tubes are also available for the D-Torch.
Tailor-made consumables for greater flexibility
We offer the following outer tubes for D-Torches:
  • Ceramic outer tube for Duo D-Torches
  • Ceramic outer tube for Radial D-Torches
  • Quartz outer tube for Duo D-Torches
  • Quartz outer tube for Radial D-Torches
Specifications
For Use With (Equipment)Duo D Torch - iCAP 7000
TypeQuartz Outer Tube
Unit SizeEach