Sprint Valve Kits for the iCAP™ 7600 ICP-OES System
Sprint Valve Kits for the iCAP™ 7600 ICP-OES System
Thermo Scientific™
Sprint Valve Kits for the iCAP™ 7600 ICP-OES System
Convenient Thermo ScientificTM Sprint Valve Kits contain all the consumables required to set up Sprint analysis for ICP-OES.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
842312052631Aqueous Sprint Valve Kit
842312052641Organics Sprint Valve Kit
Catalog number 842312052631
Price (INR)
-
Request A Quote
Type:
Aqueous Sprint Valve Kit
Enable extremely fast analysis times, utilizing the integrated Sprint Valve kits for the Thermo Scientific™ iCAP™ 7600 ICP-OES system, in conjunction with the Sprint analysis mode. These two, convenient Sprint Valve Kits contain all the consumables required to set up Sprint analysis for either aqueous or organic sample matrices.
Select Customized Consumables Kits for Easier Analysis
  • Aqueous sprint valve kit
  • Organics sprint valve kit
Specifications
For Use With (Equipment)iCAP 7600 ICP-OES System
TypeAqueous Sprint Valve Kit
Unit SizeEach