Consumable Parts for Integrated Hydride Kit for iCAP™ ICP-OES Systems
Consumable Parts for Integrated Hydride Kit for iCAP™ ICP-OES Systems
Actual product may vary
Thermo Scientific™
Consumable Parts for Integrated Hydride Kit for iCAP™ ICP-OES Systems
Ensure peak performance of your ICP-OES analyzer with dedicated consumables for analysis of hydride-generating elements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
842312052661Mini Pump PeristalticTubing
842312052611Peristaltic Pump Tubing
842318050151Hydride Kit Consumables
Catalog number 842312052661
Price (INR)
-
Request A Quote
Type:
Mini Pump PeristalticTubing
Ensure peak performance of your Thermo Scientific™ iCAP™ Series ICP-OES system using dedicated Thermo Scientific™ Consumable Parts for the Integrated Hydride Kit for ICP-OES systems.
Peak Performance with Dedicated Consumables
The replacement pump tubing and a consumables kit ensures the peak performance of hydride analysis on your iCAP Series ICP-OES System.
Consumables kit contains:
  • Pump tubing (for both, mini (95 mm) and standard peristaltic pump (152 mm))
  • Teflon membrane
  • O-ring
  • Replacement tubing
  • Sample probe
Specifications
For Use With (Equipment)Integrated Hydride Kit for ICP-OES Systems
TypeMini Pump PeristalticTubing
Unit SizeEach