Product Image
Thermo Scientific™
CETAC™ Autosampler Sample Trays for CETAC EXR-8 Autosampler
Ribs located on the bottom of these Thermo Scientific™ CETAC™ Autosampler Sample Trays hold the sample vial racks securely in place.
Have Questions?
Catalog number 842347003971
Price (INR)
-
High quality, dependable Thermo Scientific™ CETAC™ autosampler accessories make lab work easier and more productive. CETAC™ Autosampler Sample Trays feature ribs on the bottom to hold the sample vial racks securely in place.
Match Your CETAC Sample Tube Tray to Your Laboratory's Needs
  • Sample tray for CETAC ASX-260 autosampler has 10 rear standard positions and accommodates up to two sample vial racks
  • Sample tray for CETAC EXR-8 autosampler has 10 rear standard positions and accommodates up to eight sample vial racks
Specifications
TypeAutosampler Sample Tray
DescriptionTray for CETAC EXR-8 Autosampler
For Use With (Equipment)CETAC Autosampler
Unit SizeEach