Product Image
Thermo Scientific™
Power Cord with C/13 Connector, Australia
Have Questions?
Catalog NumberQuantity
848571 Cord
Catalog number 84857
Price (INR)
10,485.00
1 cord
Quantity:
1 Cord
Price (INR)
10,485.00
1 cord
This electrical power cord has a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets used in Australia.
This black, 2.4- to 3.5m-long, cord has been validated to replace the cordset that is supplied with particular Thermo Scientific powered devices. Specifically, these instruments include the myECL Imager (Part No. 62236) and the Pierce G2 Fast Blotter (Part No. 62288).
Related Products
myECL™ Imager
Pierce™ Power Station
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Cord
Unit Size1 cord