NERL™ Blood Bank Saline
Actual product may vary
NERL™ Blood Bank Saline
NERL™ Blood Bank Saline
Thermo Scientific™

NERL™ Blood Bank Saline

Enhance weak agglutination reactions with this saline, buffered to a pH of 7.0 -7.2 to match the pH of human blood.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
85041 x 20 L
85051 x 10 L
Catalog number 8504
Price (INR)
-
Quantity:
1 x 20 L
Enhance weak agglutination reactions with Thermo Scientific™ NERL™ Blood Bank Saline, buffered to a pH of 7.0 -7.2 to match the pH of human blood. Saline is available in Cubitainer™ containers with spigots for ease of use.
  • Phosphate buffered
  • Preservative-free
  • For in vitro diagnostics and laboratory use only, in immunohematological procedure or as a general laboratory diluent when “normal” or isotonic saline is indicated
  • Not for infusion, injection or irrigation

General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.

For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCertificates of Analysis are available online for this product
For Use With (Application)Phosphate-buffered saline intended for use in immunohematological procedure or as a general laboratory diluent when normal or isotonic saline is indicated.
Quantity1 x 20 L
Stability12 Months
TypeSaline, Buffered
Unit SizeEach
Contents & Storage
15°C to 30°C