Replacement Fixed Angle Rotor
Thermo Scientific™
Replacement Fixed Angle Rotor
Use this replacement fixed angle rotor, which fits a personal microcentrifuge.
Have Questions?
Catalog number 85430279
Price (INR)
-
Use the Thermo Scientific™ Replacement Fixed Rotor, which fits the Personal Microcentrifuge.
Specifications:
  • Capacity: 12 × 2mL
  • Max. Speed/RCF: 14,600rpm/14,500 x g
Compatible with:
Thermo Scientific Sorvall™ Legend™ 14 and Espresso Personal Microcentrifuges
Specifications
DescriptionReplacement Fixed Angle Rotor
For Use With (Equipment)Thermo Scientific Sorvall Legend 14 and Espresso Microcentrifuges
Capacity12 x 2 mL
Max. RCF14,500 x g
Max. Speed14,600 rpm
TypeFixed Angle Rotor
Unit SizeEach