Product Image
Invitrogen™
CellVue™ Maroon Cell Labeling Kit
CellVue™ dyes are lipophilic dyes that can be used to label the cell membrane for the purpose of identifying andRead more
Have Questions?
Catalog NumberQuantity
88-0870-161 kit
Catalog number 88-0870-16
Price (INR)
31,536.00
Online offer
Ends: 31-Mar-2026
48,516.00
Save 16,980.00 (35%)
Each
Quantity:
1 kit
Price (INR)
31,536.00
Online offer
Ends: 31-Mar-2026
48,516.00
Save 16,980.00 (35%)
Each
CellVue™ dyes are lipophilic dyes that can be used to label the cell membrane for the purpose of identifying and tracking labeled cells. Cell labeling is rapid and stable and can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy. The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C).
CellVue™ Maroon is a far-red fluorescent cell labeling reagent. It is optimally excited at 647 nm and has a peak emission of 667 nm; however, it can be detected equally in filter sets designed for APC, Alexa Fluor™ 700, and APC-eFluor™ 780 or similar fluorochromes. Therefore, using antibodies conjugated to these fluorochromes in combination with using CellVue™ Maroon is not recommended.
Reported Application
Microscopy, Cell Labeling, Immunocytochemistry, Flow Cytometric Analysis
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Reagent TypeCell Tracker Compounds, Cell Labeling Reagents
Label TypeeFluor
Product TypeCell Labeling
Unit SizeEach