Product Image
Invitrogen™
CellVue™ Plum Cell Labeling Kit
CellVue™ dyes are lipophilic dyes that can be used to label the cell membrane for the purpose of identifying andRead more
Have Questions?
Catalog NumberQuantity
88-0871-161 kit
Catalog number 88-0871-16
Price (INR)
49,885.00
Each
Quantity:
1 kit
Price (INR)
49,885.00
Each
CellVue™ dyes are lipophilic dyes that can be used to label the cell membrane for the purpose of identifying and tracking labeled cells. Cell labeling is rapid and stable and can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy. The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C).
CellVue™ Plum is a far-red fluorescent cell labeling reagent. It is optimally excited at 652 nm and has a peak emission of 671 nm, however, it can be detected equally in filter sets designed for APC, Alexa Fluor™ 700, and APC-eFluor™ 780 or similar fluorochromes. Therefore, it is not recommended to use antibodies conjugated to these fluorochromes when using CellVue™ Plum.
Reported Application
Flow Cytometric Analysis, Microscopy, Immunocytochemistry, Cell Labeling
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Reagent TypeCell Tracker Compounds, Cell Labeling Reagents
Label TypeeFluor
Product TypeCell Labeling
Unit SizeEach