CellVue™ Lavender Cell Labeling Kit
Product Image
Invitrogen™
CellVue™ Lavender Cell Labeling Kit
CellVue™ dyes are lipophilic dyes that can be used to label the cell membrane for the purpose of identifying andRead more
Have Questions?
Catalog NumberQuantity
88-0873-161 kit
Catalog number 88-0873-16
Price (INR)
21,588.00
Online offer
Ends: 31-Mar-2026
33,211.00
Save 11,623.00 (35%)
Each
Add to cart
Quantity:
1 kit
Price (INR)
21,588.00
Online offer
Ends: 31-Mar-2026
33,211.00
Save 11,623.00 (35%)
Each
Add to cart
CellVue™ dyes are lipophilic dyes that can be used to label the cell membrane for the purpose of identifying and tracking labeled cells. Cell labeling is rapid and stable and can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy. Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C).
CellVue™ Lavender is a violet fluorescent cell labeling reagent. It is optimally excited at 420 nm and has a peak emission of 461 nm. CellVue™ Lavender is compatible with most multi-color flow cytometric applications; however, due to similar emission properties it cannot be used in combination with eFluor™ 450 or Pacific Blue™ reagents.
Reported Application
Flow Cytometric Analysis, Microscopy, Immunocytochemistry, Cell Labeling
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Reagent TypeCell Tracker Compounds, Cell Labeling Reagents
Product TypeCell Labeling
Unit SizeEach