Product Image
Invitrogen™
CellVue™ Jade Cell Labeling Kit
CellVue™ dyes are lipophilic dyes that can be used to label the cell membrane for the purpose of identifying andRead more
Have Questions?
Catalog NumberQuantity
88-0876-161 kit
Catalog number 88-0876-16
Price (INR)
31,536.00
Online offer
Ends: 31-Mar-2026
48,516.00
Save 16,980.00 (35%)
Each
Quantity:
1 kit
Price (INR)
31,536.00
Online offer
Ends: 31-Mar-2026
48,516.00
Save 16,980.00 (35%)
Each
CellVue™ dyes are lipophilic dyes that can be used to label the cell membrane for the purpose of identifying and tracking labeled cells. Cell labeling is rapid and stable and can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy. Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C).
CellVue™ Jade is a green fluorescent cell labeling reagent. It is optimally excited at 478 nm and has a peak emission of 508 nm. CellVue™ Jade is compatible with most multi-color applications; however, due to similar emission properties it cannot be used in combination with FITC or Alexa Fluor™ 488 reagents.
Reported Application
Flow Cytometric Analysis, Microscopy, Immunocytochemistry, Cell Labeling
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Reagent TypeCell Tracker Compounds, Cell Labeling Reagents
Product TypeCell Labeling
Unit SizeEach