Product Image
Invitrogen™

MagniSort™ Mouse CD335 (NKp46) Positive Selection Kit

The MagniSort™ Mouse CD335 (NKp46) Positive Selection Kit is designed for the magnetic separation of CD335+ cells by positive selection.Read more
Have Questions?
Catalog NumberQuantity
8802-6817-74200 tests
Catalog number 8802-6817-74
Price (INR)
-
Quantity:
200 tests

The MagniSort™ Mouse CD335 (NKp46) Positive Selection Kit is designed for the magnetic separation of CD335+ cells by positive selection. It has been optimized for the isolation of CD335+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD335+ antibody and streptavidin-coated magnetic beads. CD335+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD335+ cells by decanting.

After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD49b (Integrin alpha 2), clone DX5, or Anti-Mouse NK1.1, clone PK136, in C57BL, FVB/N and NZB mice.

Reported Application: Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD335+
FormatKit
No. of Tests200 Tests
Quantity200 tests
Target SpeciesMouse
For Use With (Application)Magnetic Cell Separation
Product TypePositive Selection Kit
Unit SizeEach