Product Image
Invitrogen™
MagniSort™ Mouse CD117 (c-Kit) Positive Selection Kit
The MagniSort™ Mouse CD117 (c-Kit) Positive Selection Kit is designed for the magnetic separation of CD117+ cells by positive selection.Read more
Have Questions?
Catalog NumberQuantity
8802-6838-74200 tests
Catalog number 8802-6838-74
Price (INR)
-
Quantity:
200 tests
The MagniSort™ Mouse CD117 (c-Kit) Positive Selection Kit is designed for the magnetic separation of CD117+ cells by positive selection. It has been optimized for the isolation of CD117+ cells from mouse bone marrow utilizing a biotinylated Anti-Mouse CD117 antibody and streptavidin-coated magnetic beads. CD117+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD117+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD117, clone 2B8.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD117+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypePositive Selection Kit
Unit SizeEach