Product Image
Invitrogen™
MagniSort™ Mouse CD8 Positive Selection Kit
The MagniSort™ Mouse CD8 Positive Selection Kit is designed for the magnetic separation of CD8+ cells by positive selection. ItRead more
Have Questions?
Catalog NumberQuantity
8802-6842-74200 tests
Catalog number 8802-6842-74
Price (INR)
-
Quantity:
200 tests
The MagniSort™ Mouse CD8 Positive Selection Kit is designed for the magnetic separation of CD8+ cells by positive selection. It has been optimized for the isolation of CD8+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD8a antibody and streptavidin-coated magnetic beads. CD8+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD8+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD8a, clone 53-6.7.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD8+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypePositive Selection Kit
Unit SizeEach