Product Image
Invitrogen™

MagniSort™ Mouse CD45R (B220) Positive Selection Kit

The MagniSort™ Mouse CD45R (B220) Positive Selection Kit is designed for the magnetic separation of CD45R+ cells by positive selection.Read more
Have Questions?
Catalog NumberQuantity
8802-6846-74200 tests
Catalog number 8802-6846-74
Price (INR)
-
Quantity:
200 tests

The MagniSort™ Mouse CD45R (B220) Positive Selection Kit is designed for the magnetic separation of CD45R+ cells by positive selection. It has been optimized for the isolation of CD45R+ cells from mouse spleens, bone marrow, or lymph nodes utilizing a biotinylated Anti-Mouse CD45R antibody and streptavidin-coated magnetic beads. CD45R+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD45R+ cells by decanting.

After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD19, clone eBio1D3 (1D3).

Reported Application: Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD45R
FormatFormat
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
For Use With (Application)Magnetic Cell Separation
Product TypePositive Selection Kit
Unit SizeEach