Product Image
Invitrogen™
MagniSort™ Mouse CD11b Positive Selection Kit
The MagniSort™ Mouse CD11b Positive Selection Kit is designed for the magnetic separation of CD11b+ cells by positive selection. ItRead more
Have Questions?
Catalog NumberQuantity
8802-6860-74200 tests
Catalog number 8802-6860-74
Price (INR)
53,879.00
線上優惠
Ends: 31-Mar-2026
82,890.00
Save 29,011.00 (35%)
Each
Quantity:
200 tests
Price (INR)
53,879.00
線上優惠
Ends: 31-Mar-2026
82,890.00
Save 29,011.00 (35%)
Each
The MagniSort™ Mouse CD11b Positive Selection Kit is designed for the magnetic separation of CD11b+ cells by positive selection. It has been optimized for the isolation of CD11b+ cells from mouse bone marrow cells or splenocytes utilizing a biotinylated Anti-Mouse CD11b antibody and streptavidin-coated magnetic beads. Please refer to the MagniSort™ Mouse CD11b Positive Selection Protocol for specific optimal conditions for isolating CD11b+ cells from either of these two tissues. CD11b+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD11b+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD11b, clone M1/70.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD11b+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypePositive Selection Kit
Unit SizeEach