Product Image
Invitrogen™
MagniSort™ Mouse CD49b Positive Selection Kit
The MagniSort™ Mouse CD49b Positive Selection Kit is designed for the magnetic separation of CD49b+ cells by positive selection. ItRead more
Have Questions?
Catalog NumberQuantity
8802-6862-74200 tests
Catalog number 8802-6862-74
Price (INR)
-
Quantity:
200 tests
The MagniSort™ Mouse CD49b Positive Selection Kit is designed for the magnetic separation of CD49b+ cells by positive selection. It has been optimized for the isolation of CD49b+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD49b antibody and streptavidin-coated magnetic beads. CD49b+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD49b+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD49b (integrin alpha 2), clone DX5, and Anti-Mouse NK1.1, clone PK136. For mouse strains that do not express NK1.1 antigen such as BALB/c, SJL, AKR, CBA C3H, Anti-Mouse CD335 (NKp46), clone 29A1.4, may be used.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD49b+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypePositive Selection Kit
Unit SizeEach