MagniSort™ Mouse F4/80 Positive Selection Kit
Product Image
Invitrogen™
MagniSort™ Mouse F4/80 Positive Selection Kit
The MagniSort™ Mouse F4/80 Positive Selection Kit is designed for the magnetic separation of F4/80+ cells by positive selection. ItRead more
Have Questions?
Catalog NumberQuantity
8802-6863-74200 tests
Catalog number 8802-6863-74
Price (INR)
53,879.00
キャンペーン価格
Ends: 31-Mar-2026
82,890.00
Save 29,011.00 (35%)
Each
Add to cart
Quantity:
200 tests
Price (INR)
53,879.00
キャンペーン価格
Ends: 31-Mar-2026
82,890.00
Save 29,011.00 (35%)
Each
Add to cart
The MagniSort™ Mouse F4/80 Positive Selection Kit is designed for the magnetic separation of F4/80+ cells by positive selection. It has been optimized for the isolation of F4/80+ cells from mouse bone marrow or splenocytes utilizing a biotinylated Anti-Mouse F4/80 antibody and streptavidin-coated magnetic beads. F4/80+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from F4/80+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse F4/80, clone BM8, and Anti-Mouse CD11b, clones M1/70.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeF4/80+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypePositive Selection Kit
Unit SizeEach