Product Image
Invitrogen™
MagniSort™ Human CD117 (c-Kit) Positive Selection Kit
The MagniSort™ Human CD117 (c-Kit) Positive Selection Kit is designed for the magnetic separation of CD117+ cells by positive selection.Read more
Have Questions?
Catalog NumberQuantity
8802-6870-74200 tests
Catalog number 8802-6870-74
Price (INR)
-
Quantity:
200 tests
The MagniSort™ Human CD117 (c-Kit) Positive Selection Kit is designed for the magnetic separation of CD117+ cells by positive selection. It has been optimized for the isolation of CD117+ cells from normal human peripheral blood cells. CD117+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD117+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD117, clone YB5.B8, and Anti-Human CD34, clone 4H11.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD117+
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
Product TypePositive Selection Kit
Unit SizeEach