Product Image
Invitrogen™

MagniSort™ Human NK cell Enrichment Kit

The MagniSort™ Human NK cell Enrichment Kit is designed for the magnetic separation of NK cells by negative selection.
Have Questions?
Catalog NumberQuantity
8804-6819-74200 tests
Catalog number 8804-6819-74
Price (INR)
-
Quantity:
200 tests

The MagniSort™ Human NK cell Enrichment Kit is designed for the magnetic separation of NK cells by negative selection. It has been optimized for the isolation of human NK cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells, including NKT cells, are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave NK cells untouched and free in solution.

The MagniSort™ Human NK cell Enrichment Antibody Cocktail contains the following antibodies:

  • Anti-Human CD3 Biotin
  • Anti-Human CD4 Biotin
  • Anti-Human CD14 Biotin
  • Anti-Human CD19 Biotin
  • Anti-Human CD20 Biotin
  • Anti-Human CD66b Biotin
  • Anti-Human CD123 Biotin
  • Anti-Human CD235a Biotin
  • Anti-Human HLA-DR Biotin

Reported Application: Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNK Cells
FormatKit
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
For Use With (Application)Magnetic Cell Separation
Product TypeCell Enrichment Kit
Unit SizeEach