Product Image
Invitrogen™
MagniSort™ Mouse CD8 T cell Enrichment Kit
The MagniSort™ Mouse CD8 T cell Enrichment Kit is designed for the magnetic separation of mouse CD8 T cells byRead more
Have Questions?
Catalog NumberQuantity
8804-6822-74200 tests
Catalog number 8804-6822-74
Price (INR)
49,625.00
Online offer
Ends: 31-Mar-2026
76,345.00
Save 26,720.00 (35%)
Each
Quantity:
200 tests
Price (INR)
49,625.00
Online offer
Ends: 31-Mar-2026
76,345.00
Save 26,720.00 (35%)
Each
The MagniSort™ Mouse CD8 T cell Enrichment Kit is designed for the magnetic separation of mouse CD8 T cells by negative selection. It has been optimized for the isolation of mouse CD8 T cells from spleens or lymph nodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD8 T cells untouched and free in solution.
The MagniSort™ Mouse CD8 T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD4 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD24 Biotin
Anti-Mouse CD45R (B220) Biotin
Anti-Mouse CD49b (Integrin alpha 2) Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse γδ TCR Biotin
Anti-Mouse TER-119 Biotin
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (CD8)
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypeCell Enrichment Kit
Unit SizeEach