Product Image
Invitrogen™
MagniSort™ Mouse Hematopoietic Lineage Depletion Kit
The MagniSort™ Mouse Hematopoietic Lineage Depletion Kit is designed for the magnetic depletion of mouse hematopoietic cells with committed lineagesRead more
Have Questions?
Catalog NumberQuantity
8804-6829-74200 tests
Catalog number 8804-6829-74
Price (INR)
-
Quantity:
200 tests
The MagniSort™ Mouse Hematopoietic Lineage Depletion Kit is designed for the magnetic depletion of mouse hematopoietic cells with committed lineages from mouse bone marrow. It utilizes a cocktail of antibody specific for hematopoietic lineages and streptavidin-coated magnetic beads. Lineage committed cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the lineage negative cells can be separated from lineage committed cells by decanting.
The MagniSort™ Mouse Hematopoietic Lineage Depletion Antibody Cocktail contains the following antibodies:
Anti-Mouse CD2 Biotin
Anti-Mouse CD3 Biotin
Anti-Mouse CD5 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD45R Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse TER-119 Biotin
After depletion, the enrichment of multi-potent progenitor cells can be verified by staining with Anti-Mouse CD34 (clone RAM34), and the enrichment of CD34- hematopoietic stem cells can be verified by additional staining with Anti-Mouse CD150 (clone mShad150) and Anti-Mouse CD48 (clone HM48-1).
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHematopoietic Cells
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypeCell Depletion Kit
Unit SizeEach