Search
| Catalog Number | Quantity |
|---|---|
| 8822-6856-41 | 25 tests |
This eZKine™ 4-Color Compensation Control Kit is designed for compensation setup with eZKine™ Whole Blood Intracellular Cytokine Kits. It contains the necessary components to create single-color compensation controls for use with eZKine™ cocktails, including OneComp eBeads and 4 vials of antibody representing each of the 4 fluorochromes used in eZKine™ cocktails. OneComp eBeads can be stained with each of the 4 fluorochrome-conjuated antibodies to set up single-color compensation control tubes.