Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL
Thermo Scientific™

Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL

Remove endotoxins (lipopolysaccharides or LPS) from samples before mass spectrometry using spin columns of poly(e-lysine) ligand on beaded cellulose.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8827725 Columns
882765 Columns
Catalog number 88277
Price (INR)
63,350.00
Online offer
Ends: 31-Dec-2026
105,582.40
Save 42,232.40 (40%)
Each
Quantity:
25 Columns
Request bulk or custom format
Price (INR)
63,350.00
Online offer
Ends: 31-Dec-2026
105,582.40
Save 42,232.40 (40%)
Each

Thermo Scientific Pierce High Capacity Endotoxin Removal Resin binds to reduce endotoxin levels in protein samples by ≥99% in 1 hour; with this spin cup format, protein recovery is ≥85%.

Features of High Capacity Endotoxin Removal Resin Spin Columns:

  • High capacity—bind up to 2,000,000 EU/mL† to eliminate >99% of endotoxins
  • Selective—recover ≥85% of your protein sample
  • High performance—meets FDA guidelines by reducing final EU concentration to <5 EU/mL
  • Fast—our new spin cup format enables endotoxin depletion within 1 hour
  • Clean—single-use spin columns avoid cross contamination of samples
  • Optimized—spin cups are optimized for different sample volumes

The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(ε-lysine), as an affinity ligand to selectively bind endotoxins. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers.

Requires:

  • Endotoxin-free water

Applications:

  • Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection

The presence of endotoxins in biologically derived products prepared for research, vaccination, or therapeutic use is a major concern due to the diverse and potentially harmful biological response to these molecules. Endotoxin contamination in proteins purified from bacterial cultures is due to residual lipopolysaccharides (LPS) derived from the outer cell membrane of gram-negative bacteria such as E. coli. Endotoxin-contaminated protein samples transfected into cells or injected into an animal host can initiate a strong immune response resulting in a Systemic Inflammatory Response Syndrome (SIRS) and/or sepsis. Traditional endotoxin removal resins that use polymixin B and sodium deoxycholate to clear endotoxins from a sample also have the negative side effects of neurotoxicity and acute renal tubular necrosis.

Pierce High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000-EU/mL binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/mL. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/mL. This spin cup format is a fast, single-use method to remove endotoxins from protein samples in as little as 1 hour. The spin cups use a batch format to bind and remove endotoxins while allowing for >85% protein recovery.

† One endotoxin unit/mL (EU/mL) equals approximately 0.1ng endotoxin/mL of solution.

Related Products

  • Pierce™ High Capacity Endotoxin Removal Resin (Cat. No. 88270)
  • Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.25 mL (Cat. No. 88273)
  • Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.5 mL (Cat. No. 88274)
For Research Use Only. Not for use in diagnostic procedures.

The presence of endotoxins in biologically derived products prepared for research, vaccination, or therapeutic use is a major concern due to the diverse and potentially harmful biological response to these molecules. Endotoxin contamination in proteins purified from bacterial cultures is due to residual lipopolysaccharides (LPS) derived from the outer cell membrane of gram-negative bacteria such as E. coli. Endotoxin-contaminated protein samples transfected into cells or injected into an animal host can initiate a strong immune response resulting in a Systemic Inflammatory Response Syndrome (SIRS) and/or sepsis. Traditional endotoxin removal resins that use polymixin B and sodium deoxycholate to clear endotoxins from a sample also have the negative side effects of neurotoxicity and acute renal tubular necrosis.

Our Pierce High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000-EU/mL binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/mL. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/mL. This resin is offered in a slurry format for custom packing of endotoxin removal columns for gravity flow or flow rates ranging from 10-15mL/hour. Alternatively, the spin cup format is a fast, single-use method to remove endotoxins from protein samples in as little as 1 hour. The spin cups use a batch format to bind and remove endotoxins while allowing for at least 85% protein recovery and are available in 3 sizes to process protein samples of different volumes.

Specifications
Column TypeAffinity, Cellulose Resin
FormatSpin Column
Formulation22 mL-capacity centrifuge columns containing 1 mL resin
Quantity25 Columns
Stationary PhaseProprietary Ligand
TargetEndotoxin Removal
Capacity (Metric)Up to 2,000,000 EU/mL
FormLiquid
Product LinePierce
TypeSpin Column
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.

Frequently asked questions (FAQs)

Are Pierce High Capacity Endotoxin Removal Spin Columns, 1 mL (Cat. No. 88276) autoclavable?

We have not tested this, but the resin is autoclavable up to 121 degrees C at 1 bar for 20 min.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
Unraveling function and diversity of bacterial lectins in the human microbiome.
Authors:Brady SF
Journal:Nature communications
PubMed ID:35661736