Melon™ Gel Chromatography Cartridges
Actual product may vary
Melon™ Gel Chromatography Cartridges
Melon™ Gel Chromatography Cartridges
Melon™ Gel Chromatography Cartridges
Thermo Scientific™
Melon™ Gel Chromatography Cartridges
The Thermo Scientific Melon Gel Chromatography Cartridge is a convenient, ready-to-use prepacked chromatography column for the isolation and purification ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
899331 cartridge x 5 mL
899322 x 1 mL
Catalog number 89933
Price (INR)
65,803.00
Each
Quantity:
1 cartridge x 5 mL
Request bulk or custom format
Price (INR)
65,803.00
Each
The Thermo Scientific Melon Gel Chromatography Cartridge is a convenient, ready-to-use prepacked chromatography column for the isolation and purification of antibodies. Our Melon Gel IgG Purification Resin offers a simpler and faster alternative to traditional Protein A and Protein G methods for purification of human, mouse and rabbit IgG from serum and other sources.
Features of Melon Gel IgG Purification Resin:
Simple, one-step protocol—IgG purification with Melon Gel Resin does not involve binding, washing, and multiple elution steps required by Protein A and Protein G
Rapid purification—antibody purification from serum is four to six times faster than Protein A or G methods
High recovery and purity—for many species of serum antibody recovery is greater than 90% with purity greater than 80%
Minimal dilution—reduced need to concentrate antibody before storage or use
Gentle purification—No harsh antibody elution conditions helps retain IgG activity
Regenerable support—Melon Gel Resin can be used for multiple antibody purifications
These cartridges are compatible with major automated liquid-chromatography systems or manual syringe processing, and attach directly to ÄKTA™ or FPLC Systems without additional connectors. An accessory pack, included with each product, readily adapts cartridges for use with Luer-Lok Syringe Fittings or 1/16” tubing. The Melon Gel Cartridges provide fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
Melon Gel IgG Purification Resin enables purification of antibodies from serum and other sample types. Melon Gel Resin binds and removes serum proteins, thereby allowing pure IgG to be collected in the flow-through fraction. The procedure is gentle because no harsh elution conditions are involved.
Melon Gel Resin contains a proprietary ligand that retains most protein found in serum, ascites and culture supernatants, while allowing IgG to pass through the support and be collected in the flow-through fraction. Because Melon Gel Resin is not a bind-and-release support, it is extremely fast and gentle to ant ibodies, resulting in antibody preparations of high purity and high activity.
Melon Gel Resin works with antibodies from a variety of species and subclasses, many of which do not purify efficiently with Protein A or Protein G. The resulting recovery and purity of the IgG isolated by this method rivals that obtained from the same samples using bind-and-release supports such as Protein A or Protein G. Furthermore, antibodies purified with Melon Gel Resin are recovered ready for use in immunoassays or subsequent conjugation or modification steps; there is no need to neutralize or desalt the recovered antibody to remove harsh elution buffers.
Melon Gel IgG Purification saves time compared to classical Protein A or Protein G methods without compromising the quality of the antibody. Compared to antibody affinity purification with Protein A or Protein G, Melon Gel IgG Purification is simpler, faster and provides greater yields and higher purity for a wider variety of antibodies species.
Related Products
Melon™ Gel Purification Buffer
Melon™ Gel Regenerant
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Flow Rate3 mL/min.
Purification TargetIgG Antibodies
Quantity1 cartridge x 5 mL
Volume (Metric) Bed5 mL
Column TypeProprietary Resin, Affinity
FormatChromatography Cartridge
Product LineMelon
Product TypeChromatography Cartridge
Stationary PhaseProprietary Ligand
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C to 8°C. Do not freeze.
Frequently asked questions (FAQs)
How can I determine when the Melon Gel Support is saturated?
For serum, a volume double the Melon Gel Support volume is the maximum amount that can be processed at one time. To determine if too much sample was processed, examine the purified IgG by SDS-PAGE for additional protein bands.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can Melon Gel purify chicken IgY?
No. Thermo Scientific Thiophilic Adsorbent (Cat. Nos. 20500 and 44916) can be used to purify IgY from serum. The Chicken IgY Purification Kit (Cat. No. 44918) can be used to purify IgY from egg yolks.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Is the Melon Gel IgG Spin Purification Kit (Cat. No. 45206) a viable alternative to using traditional Protein A and Protein G methods for purification of IgG from serum?
Yes. With the Melon Gel IgG Spin Purification Kit, 90% recovery of IgG with greater than 80% purity are typically observed for a single purification. Although IgA and IgM may be present along with the purified IgG, a high percentage (> 80%) of IgA and IgM is retained on the gel support. This high binding of IgA and IgM to the gel support combined with the fact that IgA and IgM are present in low abundance in serum results in low levels of these antibody molecules in the purified IgG. Here are other advantages of using the Melon Gel IgG Spin Purification Kit over traditional Protein A or Protein G methods:
-Simple, one-step protocol - IgG purification with Melon Gel Resin does not involve binding, washing, and multiple elution steps required by Protein A and Protein G
-Rapid purification - antibody purification from serum is four to six times faster than Protein A or G methods
-Minimal dilution - reduced need to concentrate antibody before storage or use
-Robust purification - Melon Gel IgG Purification Kits work with a wide range of antibodies including many that do not purify well on Protein A or Protein G
-Gentle purification - No harsh antibody elution conditions helps retain IgG activity
-Regenerable support - Melon Gel Resin can be used for multiple antibody purifications
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How many times can I reuse the Melon Gel Support?
The Melon Gel Support can be used up to three times. Performing an ammonium sulfate precipitation on serum samples containing large amounts of non-antibody protein before purification improves purification efficiency and increases reusability.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
After Melon Gel purification, must I desalt or dialyze the purified antibody before using the Thermo Scientific Direct IP Kit (Cat. No. 26148)?
No. However, researchers may want to add an equal volume of binding buffer to elevate the pH towards neutral.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.