Product Image
Applied Biosystems™
Probe Array Cartridge Carriers (Red)
Have Questions?
Catalog NumberQuantity
90-03561 each
Catalog number 90-0356
Price (INR)
11,645.00
Each
Quantity:
1 each
Price (INR)
11,645.00
Each
Probe Array Cartridge Carriers (Red)
Cartridge carriers, designed specifically for use with the GeneChip Hybridization Oven 640 and 645, are molded from advanced composite materials and hold eight GeneChip probe array cartridges per carrier. Carriers have barcodes for tracking purposes.
GeneChip Hybridization Oven 645
The GeneChip Hybridization Oven 645 provides precise temperature and rotation control to ensure the successful hybridization of as many as 64 GeneChip cartridges at one time.
An ergonomic front-panel display contains system status indicators and buttons for adjusting the set point temperature and the rotisserie speed. This panel, in addition to a highly visible display and large window, provides immediate feedback of proper operation when using the oven.
The oven can be configured for 110 or 220V operation.
For more information, please review the data sheet (pdf, 126 KB).
Related Links
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)GeneChip Hybridization Oven
Quantity1 each
ColorRed
Product TypeProbe Array Cartridge Carrier
Unit SizeEach
Frequently asked questions (FAQs)
Which probe array cartridge carriers are designed specifically for use with the GeneChip Hybridization Oven 645 (Cat. No. 00-0331)?
The following probe array cartridge carriers are designed for use with the GeneChip Hybridization Oven 645:
- Probe Array Cartridge Carriers (Red) (Cat. No. 90-0356)
- Probe Array Cartridge Carriers (Green) (Cat. No. 90-0357)
- Probe Array Cartridge Carriers (Purple) (Cat. No. 90-0358)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.