ABTS ELISA Buffer Kit, PeproTech®
Invitrogen™
ABTS ELISA Buffer Kit, PeproTech®
This kit contains the key components required for the analysis of ten sandwich ELISA plates. It has been specifically formulatedRead more
Have Questions?
Catalog NumberQuantity
900-K001 Kit
Catalog number 900-K00
Price (INR)
23,284.00
Each
Quantity:
1 Kit
Price (INR)
23,284.00
Each
This kit contains the key components required for the analysis of ten sandwich ELISA plates. It has been specifically formulated to be used in conjunction with PeproTech standard ELISA development kits.
Included solutions and materials:
• PBS
• Wash buffer
• Blocking buffer
• Diluent
• ABTS liquid substrate
• ELISA microplates
• Sealing film
Specifications
Detection MethodColorimetric
For Use With (Application)ELISA
Product LinePeproTech
Substrate TypeABTS, HRP
Product TypeELISA Buffer
Quantity1 Kit
Unit SizeEach