TMB ELISA Buffer Kit, PeproTech®
Invitrogen™
TMB ELISA Buffer Kit, PeproTech®
This kit contains the key components required for the analysis of ten sandwich ELISA plates. It has been specifically formulatedRead more
Have Questions?
Catalog NumberQuantity
900-T001 Kit
Catalog number 900-T00
Price (INR)
23,284.00
Each
Quantity:
1 Kit
Price (INR)
23,284.00
Each
This kit contains the key components required for the analysis of ten sandwich ELISA plates. It has been specifically formulated to be used in conjunction with PeproTech standard TMB ELISA development kits.
Included solutions and materials:
• PBS
• Wash buffer
• Blocking buffer
• Diluent
• TMB liquid substrate
• ELISA microplates
• Sealing film
• Stop solution
Specifications
Detection MethodColorimetric
For Use With (Application)ELISA
Product LinePeproTech
Substrate TypeTMB, HRP
Product TypeELISA Buffer
Quantity1 Kit
Unit SizeEach