Product Image
Applied Biosystems™

Attune™ Flow Cytometer Wash Bottle

This is a 250 mL replacement bottle for wash solution for the Attune Flow Cytometer.Read more
Have Questions?
Catalog NumberQuantity
90032053250 mL
Catalog number 90032053
Price (INR)
-
Quantity:
250 mL
This is a 250 mL replacement bottle for wash solution for the Attune Flow Cytometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune
Quantity250 mL
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeWash Bottle
Unit SizeEach
Contents & Storage
Store at room temperature