GeneChip™ Mouse Genome 430A 2.0 Array
Applied Biosystems™
GeneChip™ Mouse Genome 430A 2.0 Array
The GeneChip™ Mouse Genome 430A 2.0 Array is a single array representing approximately 14,000 well-characterized mouse genes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
900500TS
also known as 900500
30 arrays
9004982 arrays
9004996 arrays
Catalog number 900500TS
also known as 900500
Price (INR)
1,902,091.00
Each
Number of Arrays:
30 arrays
Price (INR)
1,902,091.00
Each
The GeneChip™ Mouse Genome 430A 2.0 Array is a single array representing approximately 14,000 well-characterized mouse genes that can be used to explore biology and disease processes. New, reduced feature size means that you can use smaller sample volumes than the previous Mouse Expression Array 430A without compromising performance.
  • Provides coverage of well-substantiated genes in the transcribed mouse genome on a single array
  • Analyzes the expression level of approximately 14,000 well-characterized mouse genes
  • The Power of the Probe Set - offering multiple independent measurements for each transcript delivering the greatest accuracy and reproducibility of any microarray platform
  • All probe sets represented on the GeneChip™ Mouse Expression Array 430A are included on the GeneChip Mouse Genome 430A 2.0 Array
Array Profile
Sequences used in the design of the array were selected from GenBank™, dbEST, and RefSeq. The sequence clusters were created from the UniGene database (Build 107, June 2002) and then refined by analysis and comparison with the publicly available draft assembly of the mouse genome from the Whitehead Institute for Genome Research (MGSC, April 2002).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity30 arrays
TypeConnexin Antibody Sample Pack
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays30 arrays
SpeciesMouse
Unit SizeEach