GeneChip™ Rat Genome 230 2.0 Array
Applied Biosystems™
GeneChip™ Rat Genome 230 2.0 Array
The GeneChip™ Rat Genome 230 2.0 Array is a powerful tool for toxicology, neurobiology, and other applications using rat asRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
90050730 arrays
9005052 arrays
9005066 arrays
Catalog number 900507
Price (INR)
-
Number of Arrays:
30 arrays
The GeneChip™ Rat Genome 230 2.0 Array is a powerful tool for toxicology, neurobiology, and other applications using rat as a model organism.
• Provides comprehensive coverage of the transcribed rat genome on a single array
• Comprised of more than 31,000 probe sets, analyzing over 30,000 transcripts and variants from over 28,000 well-substantiated rat genes
• Sophisticated bioinformatics tools available through the NetAffx™ Analysis Center to rapidly obtain biologically meaningful results
• Publicly available drafts of the rat genome and leading public rat databases were used to refine sequences and provide a higher quality of data output
Array Profile
All probe sets represented on the GeneChip Rat Expression Set 230 are included on the GeneChip Rat Genome 230 2.0 Array. Sequences used in the design of the GeneChip Rat Genome 230 2.0 Array were selected from GenBank™, dbEST, and RefSeq. The sequence clusters were created from the UniGene database (Build 99, June 2002) and then refined by analysis and comparison with the publicly available draft assembly of the rat genome from the Baylor College of Medicine Human Genome Sequencing Center (June 2002).
The GeneChip Rat Genome 230 2.0 Array includes representation of the RefSeq database sequences and probe sets related to sequences and refined EST clusters previously represented on the GeneChip Rat Genome U34 Set.
Oligonucleotide probes complementary to each corresponding sequence are synthesized in situ on the arrays. Eleven pairs of oligonucleotide probes are used to measure the level of transcription of each sequence represented on the GeneChip Rat Genome 230 2.0 Array.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity30 arrays
TypeRat Genome 230 2.0 Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays30 arrays
SpeciesRat
Unit SizeEach
Frequently asked questions (FAQs)
What reagent kit should I use with my array?
Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How are transcript IDs assigned for WT arrays if a gene has multiple transcript IDs?
The first assigned mRNA/gene that is listed in the annotation file for a given probe set or transcript cluster is considered the "best" assignment based on data source quality ranking and assignment scoring system. To get a single best mRNA/gene assignment you can simply take the first one in the list in the annotation file.
Sometimes, there might be multiple best hits between transcript clusters (or probe set) and a mRNA/gene, with identical score and data source quality. In that case, the transcript assignments are ranked based on their descriptions (e.g., title from the GenBank record).
For transcript clusters that detect genes that are alternatively spliced, the different spliced forms will be ranked by length with the longest ones listed first.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Does the GeneChip miRNA 3.1 Array Strip require a different hyb/wash/stain kit than the GeneChip 3' IVT arrays and GeneChip ST array strips?
The hyb/wash/stain kit for the GeneChip miRNA 3.1 Array Strips is different than that for the 3' IVT arrays and GeneChip ST array strips. Additional wash A steps have been added to ensure adequate washing after the array strips have been stained.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What do the suffixes mean at the end of each probeset in GeneChip 3' IVT arrays?
The suffixes, at the end of each probeset in GeneChip 3' IVT arrays, indicate what is known about the sequence at the time of design:
_at = all the probes hit one known transcript
_a = all probes in the set hit alternate transcripts from the same gene
_s = all probes in the set hit transcripts from the different genes
_x = some probes hit transcripts from different genes
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I store my GeneChip 3' IVT Array after hybridization?
Yes, but we recommend storing your GeneChip 3' IVT Array after hybridization and prior to the fluidics step, for no longer than 3 hours at 4 degrees C. Please remove the hybridization cocktail and fill with wash A before storing.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.