GeneChip™ S. aureus Genome Array
Applied Biosystems™
GeneChip™ S. aureus Genome Array
This GeneChip™ array is available through our Made-to-Order Program, which enables you to purchase selected custom array designs and previous-generation GeneChip arrays no longer available as catalog products.
Have Questions?
Catalog NumberQuantity
9005146 arrays
Catalog number 900514
Price (INR)
-
Quantity:
6 arrays
This GeneChip™ array is available through our Made-to-Order Program, which enables you to purchase selected custom array designs and previous-generation GeneChip arrays no longer available as catalog products.
For comprehensive monitoring of the relative mRNA abundance of S. aureus sequences.
The GeneChip S. aureus Genome Array is useful for studying the expression of sequences in the following four strains of Staphylococcus aureus: N315 (National Institute of Technology and Evaluation, Japan), Mu50 (National Institute of Technology and Evaluation, Japan), NCTC 8325 (University of Oklahoma), and COL (TIGR).
The array contains probe sets to over 3,300 S. aureus open reading frames. Additionally, the array also contains probes to study both forward and reverse orientation of over 4,800 intergenic regions throughout the S. aureus genome.
Related Links:
  • Learn more about ordering and support of GeneChip Custom Arrays.
  • For more information, please review the data sheet (pdf, 114 KB).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity6 arrays
TypeS. aureus Genome Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays6 arrays
SpeciesS. aureus
Unit Size6 arrays
Frequently asked questions (FAQs)
How should I prepare my hybridization cocktail for the GeneChip S. aureus array?
If using the IVT Pico Assay to generate labeled material, please contact the technical support team at techsupport@thermofisher.com for additional information on hybridization specifics.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do I process S. aureus samples for expression analysis on the GeneChip S. aureus array?
S. aureus samples can be successfully processed for expression analysis by following the IVT Pico Assay with modifications. Please contact the technical support team at techsupport@thermofisher.com for additional information.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.