GeneChip™ Soybean Genome Array
Applied Biosystems™
GeneChip™ Soybean Genome Array
Have Questions?
Catalog NumberQuantity
9005252 arrays
Catalog number 900525
Price (INR)
-
Quantity:
2 arrays
The GeneChip™ Soybean Genome Array can be used to study gene expression of over 37,500 soybean (Glycine max) transcripts.
The array also contains transcripts for studying two pathogens important for soybean research. Specifically, the array includes probe sets to detect approximately 15,800 transcripts for Phytophthora sojae (a water mold that commonly attacks soybean crops) as well as 7,500 Heterodera glycines (cyst nematode pathogen) transcripts.
The Soybean Genome Array was designed in close collaboration with the Soybean Research Community as part of the GeneChip Consortia Program.
Learn more about the microarray through the GeneChip Soybean Genome Array data sheet.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity2 arrays
TypeSoybean Genome Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays2 arrays
SpeciesSoybean
Unit Size2 arrays