GeneChip™ Citrus Genome Array
Applied Biosystems™
GeneChip™ Citrus Genome Array
The GeneChip™ Citrus Genome Array is designed specifically to monitor gene expression in Citrus. This array was created in collaboration with leading Citrus researchers through the Affymetrix GeneChip™ Consortia Program.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
9007312 arrays
9007326 arrays
Catalog number 900731
Price (INR)
170,415.00
2 arrays
Number of Arrays:
2 arrays
Price (INR)
170,415.00
2 arrays
The GeneChip™ Citrus Genome Array is designed specifically to monitor gene expression in Citrus. This array was created in collaboration with leading Citrus researchers through the Affymetrix GeneChip™ Consortia Program. The array contains 30,171 probe sets representing up to 33,879 Citrus transcripts based on EST sequences obtained from several Citrus species and Citrus hybrids. Because Citrus has relatively long generations and times-to-production, it will be especially useful for researchers to identify the genetic mechanisms critical to:
  • High crop yield
  • Disease and insect resistance
  • Growth response to environmental factors
The GeneChip™ Citrus Genome Array provides a tool that enables researchers to elucidate these complex genetic traits in Citrus to determine how crop production can be improved. The array includes 46 probe sets representing several Citrus pathogens, and 60 probe sets representing various reporters, selectable markers, and transgenes. Also included are sequences from Poncirus and Poncirus crosses with Citrus hybrids.
The GeneChip™ Citrus Genome Array was designed in close collaboration with the Citrus Research Community as part of the Affymetrix™ GeneChip Consortia Program.
Related Links:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity2 arrays
TypeCitrus Genome Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays2 arrays
SpeciesCitrus
Unit Size2 arrays