GeneChip™ Cotton Genome Array
Applied Biosystems™
GeneChip™ Cotton Genome Array
The GeneChip Cotton Genome Array is a single GeneChip brand array comprised of 239,777 probe sets representing 21,854 cotton transcripts.Read more
Have Questions?
Catalog NumberQuantity
9010002 arrays
Catalog number 901000
Price (INR)
124,338.00
2 arrays
Quantity:
2 arrays
Price (INR)
124,338.00
2 arrays
The GeneChip Cotton Genome Array is a single GeneChip brand array comprised of 239,777 probe sets representing 21,854 cotton transcripts.
The GeneChip™ Cotton Genome Array is a single GeneChip brand array comprised of 239,777 probe sets representing 21,854 cotton transcripts. Sequences used in the design of the array were selected from GenBank™, dbEST, and RefSeq, and were created from the Gossypium hirsutum UniGene database (Build 2, August 2006) and the Gossypium raimondii UniGene database (Build 2, September 2005). These sequences derive from the many contributors from the Cotton Community. Some select sequences were generously made publicly available prior to publication by a group within the Cotton Community specifically to enhance the design of the GeneChip Cotton Genome Array.
The GeneChip™ Cotton Genome Array is based on EST sequences from four species of Cotton: Gossypium hirsutum, Gossypium raimondii, Gossypium arboretum, and Gossypium barbadense. The Array was designed in close collaboration with the Cotton Research Community as part of the GeneChip Consortia Program.
For more information, please review the Package Insert (pdf, 209 KB).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity2 arrays
TypeCotton Genome Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays2 arrays
SpeciesCotton
Unit Size2 arrays